Merck
  • A cholesterol consensus motif is required for efficient intracellular transport and raft association of a group 2 HA from influenza virus.
A cholesterol consensus motif is required for efficient intracellular transport and raft association of a group 2 HA from influenza virus.
The Biochemical journal (2014-10-22)
Maren de Vries, Andreas Herrmann, Michael Veit
ZUSAMMENFASSUNG
The HA (haemagglutinin) of influenza viruses must be recruited to membrane rafts to perform its function in membrane fusion and virus budding. We previously showed using FRET that deletion of the two raft-targeting features of HA, S-acylation at the cytoplasmic tail and the hydrophobic amino acids VIL (Val-Ile-Leu) in the outer part of the TMR (transmembrane region), lead to reduced raft association. In addition, exchange of VIL, but not of the S-acylation sites severely retards transport of HA through the Golgi. In the present study, we have further characterized the ill-defined signal in the TMR. A sequence comparison suggests that the leucine residue of VIL might be part of a CCM (cholesterol consensus motif) that is known to bind cholesterol to seven-transmembrane receptors. The signal also comprises a lysine residue and a tryptophan residue on one and a tyrosine residue on another TMR helix and is conserved in group 2 HAs. Mutations in the CCM retard Golgi-localized processing of HA, such as acquisition of Endo H (endoglycosidase H)-resistant carbohydrates in the medial Golgi and proteolytic cleavage in the TGN (trans-Golgi network). The delay in transport of HA to and from the medial Golgi varied with the mutation, suggesting that different transport steps are affected. All mutants analysed by FRET also showed reduced association with rafts at the plasma membrane. Thus the raft-targeting signal of HA encompasses not only hydrophobic, but also aromatic and positively charged, residues. We speculate that binding to cholesterol might facilitate intracellular transport of HA and association with rafts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
DL-Methionin, ≥99%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Sigma-Aldrich
DL-Cystein, technical grade
Methionin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
DL-Methionin, ≥99.0% (NT)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
DL-Methionin, European Pharmacopoeia (EP) Reference Standard
Supelco
4-tert.-Octylphenol-monoethoxylat -Lösung, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg