Merck
Oxidative uncoupling in cysteine dioxygenase is gated by a proton-sensitive intermediate.
Biochemistry (2014-11-12)
Joshua K Crowell, Wei Li, Brad S Pierce
ZUSAMMENFASSUNG
Cysteine dioxygenase (CDO) is a non-heme mononuclear iron enzyme that catalyzes the O2-dependent oxidation of l-cysteine (Cys) to produce cysteine sulfinic acid (CSA). This enzyme catalyzes the first committed step in Cys catabolism; thus, it is central to mammalian sulfur metabolism and redox homeostasis. Ironically, despite nearly 45 years of continued research on CDO, essentially no information has been reported with respect to its kinetic mechanism. In this work, the timing of chemical steps in the CDO kinetic mechanism is investigated by pH/pD-dependent steady-state kinetics and solvent isotope effects on kcat, kcat/KM, and (O2/CSA) coupling. Normal solvent kinetic isotope effects of 1.45 ± 0.05 and 2.0 ± 0.1 are observed in kcat-pL and kcat/KM-pL profiles, respectively. Proton inventory experiments within the pL-independent region (pL 8.5) suggest multiple solvent-exchangeable protons in flight for both kcat and kcat/KM data. The influence of solvent viscosity was also investigated to probe non-chemical steps and to verify that the apparent isotope effects were not attributable to increased solvent viscosity of D2O reactions relative to H2O. Although solvent viscosity did have a modest influence on kcat and kcat/KM, the response is not sufficient to account for the observed solvent isotope effects. This suggests that product release is only partially rate-limiting for CDO catalysis. Most crucially, proton inventory of (O2/CSA) coupling indicates that a proton-sensitive transition state directly follows O2 activation. Thus, protonation of a transient species preceding Cys oxidation is gated by protons in flight. This behavior provides valuable insight into the kinetically masked transients generated during catalysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
SAFC
L-Cystein
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
L-Cystein, ≥97%, FG
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Supelco
L-Cystein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D