Merck
  • Potential biological role of laccase from the sponge Suberites domuncula as an antibacterial defense component.
Potential biological role of laccase from the sponge Suberites domuncula as an antibacterial defense component.
Biochimica et biophysica acta (2014-12-03)
Qiang Li, Xiaohong Wang, Michael Korzhev, Heinz C Schröder, Thorben Link, Muhammad Nawaz Tahir, Bärbel Diehl-Seifert, Werner E G Müller
ZUSAMMENFASSUNG
Laccases are copper-containing enzymes that catalyze the oxidation of a wide variety of phenolic substrates. We describe the first poriferan laccase from the marine demosponge Suberites domuncula. This enzyme comprises three characteristic multicopper oxidase homologous domains. Immunohistological studies revealed that the highest expression of the laccase is in the surface zone of the animals. The expression level of the laccase gene is strongly upregulated after exposure of the animals to the bacterial endotoxin lipopolysaccharide. To allow the binding of the recombinant enzyme to ferromagnetic nanoparticles, a recombinant laccase was prepared which contained in addition to the His-tag, a Glu-tag at the N-terminus of the enzyme. The recombinant laccase was enzymatically active. The apparent Michaelis constant of the enzyme is 114 μM, using syringaldazine as substrate. Exposure of E. coli to the nanoparticles, coated with Glu-tagged laccase, and to the mediator 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) in the presence of lignin, as the oxidizable substrate, resulted in an almost complete inhibition of colony formation. Quantitative studies of the effect of the laccase-coated iron oxide nanoparticles were performed using E. coli grown in suspension in reaction tubes within a magnetic nanoparticle separator. This newly designed magnetic nanoparticle separator allowed a removal of the nanoparticles after terminating the reaction. Using this system, a strong dose-dependent inhibition of the growth of E. coli by the laccase iron oxide nanoparticles was determined. From our data we conclude that the sponge laccase is involved in the anti-bacterial defense of the sponge organism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Eisen(III)-oxid, powder, <5 μm, ≥96%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Laccase aus Trametes versicolor, powder, light brown, ≥0.5 U/mg
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Eisen(III)-oxid, nanopowder, <50 nm particle size
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Eisen(III)-oxid, ≥99.995% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Laccase aus Agaricus bisporus, powder, deep brown, ≥4 U/mg
Sigma-Aldrich
Syringaldazin, indicator for laccase and peroxidase activity
Sigma-Aldrich
Eisen(III)-Oxid, Dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Syringaldazin, 98%
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri