Merck
  • The BREASTrial: stage I. Outcomes from the time of tissue expander and acellular dermal matrix placement to definitive reconstruction.
The BREASTrial: stage I. Outcomes from the time of tissue expander and acellular dermal matrix placement to definitive reconstruction.
Plastic and reconstructive surgery (2014-12-30)
Shaun D Mendenhall, Layla A Anderson, Jian Ying, Kenneth M Boucher, Ting Liu, Leigh A Neumayer, Jayant P Agarwal
ZUSAMMENFASSUNG
Use of acellular dermal matrix in tissue expander breast reconstruction has become a popular adjunct to the total submuscular technique. The question remains as to which matrix, if any, is ideal for breast reconstruction. A randomized trial was conducted to analyze outcomes of immediate staged tissue expander breast reconstruction using either AlloDerm or DermaMatrix. The impact of obesity, radiation, and chemotherapy on complications and biointegration of matrix was investigated. The trial was divided into three stages, with stage I results reported here. One hundred twenty-eight patients (199 breasts) were randomized equally over 2.5 years. Most patients were white, healthy nonsmokers. The overall complication rate was 36.2 percent; half of the complications were minor. The AlloDerm and DermaMatrix groups had similar rates of complications (33.6 percent versus 38.8 percent; p = 0.52), consisting mostly of skin necrosis (17.8 percent versus 21.4 percent; p = 0.66) and infections (13.9 percent versus 16.3 percent; p = 0.29), both of which led to tissue expander losses (5 percent versus 11.2 percent; p = 0.11). The AlloDerm group required less time for completion of expansion (42 days versus 70 days; p < 0.001). Obesity was associated with poor matrix biointegration and a longer drain time, both of which were associated with higher complication rates. The Breast Reconstruction Evaluation Using Acellular Dermal Matrix as a Sling Trial is the largest randomized trial to date in matrix breast reconstruction and emphasizes the importance of careful patient and allograft selection to minimize complications. Therapeutic, II.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder