Merck
Metabolism. Differential regulation of mTORC1 by leucine and glutamine.
Science (New York, N.Y.) (2015-01-09)
Jenna L Jewell, Young Chul Kim, Ryan C Russell, Fa-Xing Yu, Hyun Woo Park, Steven W Plouffe, Vincent S Tagliabracci, Kun-Liang Guan
ZUSAMMENFASSUNG
The mechanistic target of rapamycin (mTOR) complex 1 (mTORC1) integrates environmental and intracellular signals to regulate cell growth. Amino acids stimulate mTORC1 activation at the lysosome in a manner thought to be dependent on the Rag small guanosine triphosphatases (GTPases), the Ragulator complex, and the vacuolar H(+)-adenosine triphosphatase (v-ATPase). We report that leucine and glutamine stimulate mTORC1 by Rag GTPase-dependent and -independent mechanisms, respectively. Glutamine promoted mTORC1 translocation to the lysosome in RagA and RagB knockout cells and required the v-ATPase but not the Ragulator. Furthermore, we identified the adenosine diphosphate ribosylation factor-1 GTPase to be required for mTORC1 activation and lysosomal localization by glutamine. Our results uncover a signaling cascade to mTORC1 activation independent of the Rag GTPases and suggest that mTORC1 is differentially regulated by specific amino acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutaminlösung 200 mmol/l, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Hexokinase aus Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hexokinase aus Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Sigma-Aldrich
L-Leucin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
L-Leucin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Leucin, 99%, FG
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Leucin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland