Merck
  • A novel flow-based assay reveals discrepancies in ADAMTS-13 inhibitor assessment as compared with a conventional clinical static assay.
A novel flow-based assay reveals discrepancies in ADAMTS-13 inhibitor assessment as compared with a conventional clinical static assay.
Journal of thrombosis and haemostasis : JTH (2014-07-22)
R Grillberger, B Gruber, S Skalicky, G Schrenk, P Knöbl, B Plaimauer, P L Turecek, F Scheiflinger, H Rottensteiner
ZUSAMMENFASSUNG
Several static Bethesda-type assays are routinely used to determine ADAMTS-13-neutralizing autoantibodies in acquired thrombotic thrombocytopenic purpura (TTP), but the inhibitory activity of these antibodies has not been thoroughly evaluated under the more physiologic condition of flow. We investigated whether ADAMTS-13 inhibitor assessment with the FRETS-VWF73 assay is predictive for evaluation under flow. Anti-ADAMTS-13 autoantibodies were purified from patients with acquired TTP by chromatography involving an ADAMTS-13 affinity matrix and/or protein G. ADAMTS-13 activity was measured with the FRETS-VWF73 assay and a novel flow assay determining the ADAMTS-13-mediated decrease in platelet aggregate surface coverage, caused by perfusion of a suspension containing platelets, erythrocytes and von Willebrand factor (VWF) over a surface coated with extracellular matrix components. The neutralizing activities of ADAMTS-13 inhibitors were compared under static conditions and under flow by use of the two assays. The suitability of the flow-based ADAMTS-13 activity assay for quantification of ADAMTS-13 inhibitors could be demonstrated by reversibility of the ADAMTS-13-dependent decrease in surface coverage upon addition of goat ADAMTS-13 antiserum. Testing the neutralizing activity of purified autoantibodies from six patients in the flow assay according to their FRETS-VWF73-based inhibitor titers gave rise to vastly different inhibitory effects, indicating a discrepancy in inhibitor assessment between static and flow conditions. Anti-ADAMTS-13 autoantibodies may show inhibitory properties in vivo that are not consistent with the ADAMTS-13 inhibitor levels determined in routine static assays, possibly because certain epitopes are selectively exposed under shear. Consequently, the course of disease and treatment efficacy may vary among TTP patients, despite common inhibitor titers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Aspirin (Acetylsalicylsäure), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetylsalicylsäure, analytical standard
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)