Merck
  • Protective effects of endothelin receptor A and B inhibitors against doxorubicin-induced cardiomyopathy.
Protective effects of endothelin receptor A and B inhibitors against doxorubicin-induced cardiomyopathy.
Biochemical pharmacology (2015-02-11)
Matthias Schwebe, Sabine Ameling, Elke Hammer, Judith V Monzel, Karina Bonitz, Susanne Budde, Kathleen Schult, Stefan Oswald, Eberhard Scheuch, Markus Grube, Axel Poesch, Thomas Budde, Ralf Ewert, Henry W S Schroeder, Heyo K Kroemer, Sandra Bien-Möller
ZUSAMMENFASSUNG
The clinical efficiency of the highly potent antitumor agent doxorubicin is limited by cardiotoxic effects. In a murine doxorubicin cardiotoxicity model, increased endothelin-1 (ET-1) expression and cardioprotective effects of the dual ET-1 blocker bosentan were demonstrated. To date it is unclear if combined blocking of endothelin A/B receptors is necessary or whether selective inhibition of one of the ET-1 receptors is sufficient for the observed cardioprotection. Therefore, we investigated the impact of dual (bosentan) and single endothelin receptor antagonism through sitaxentan (receptor A blocker) or BQ788 (receptor B blocker) in a murine doxorubicin cardiotoxicity model (C57BL/6N). Simultaneous administration of each endothelin receptor antagonist (ERA) with doxorubicin resulted in a significantly improved hemodynamic performance in comparison to the impaired cardiac function in control mice with bosentan being most effective but closely followed by sitaxentan and also BQ788. This cardioprotection was not caused by diminished doxorubicin levels in heart since the doxorubicin content in cardiac tissue was not altered by ERAs significantly. However, whole transcript expression profiling showed partly different effects of the ERAs on doxorubicin-modulated cardiac gene expression of genes involved in signal transduction (e.g. Stat3, Pim1, Akt1, Plcb2), fibrosis (e.g. Myl4), energy production (e.g. Ant1) or oxidative stress (e.g. Aox1). Furthermore, doxorubicin-mediated gene regulations were verified in the murine cardiomyocyte model HL-1 showing partly reversed expression patterns after co-administration of the ERAs. In summary, our results demonstrate strong cardioprotective effects of blocking ET-1 receptors against the doxorubicin-related cardiomyopathy and provide evidence to potential underlying signaling pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture