Merck
  • One platform comparison of estrone and folic acid anchored surface engineered MWCNTs for doxorubicin delivery.
One platform comparison of estrone and folic acid anchored surface engineered MWCNTs for doxorubicin delivery.
Molecular pharmaceutics (2014-12-18)
Neelesh Kumar Mehra, N K Jain
ZUSAMMENFASSUNG
Our main aim in the present investigation was to assess and compare the in vitro and in vivo cancer targeting propensity of doxorubicin (DOX) loaded folic acid (FA) and estrone (ES) anchored PEGylated multiwalled carbon nanotubes (MWCNTs) employing tumor bearing Balb/c mice. The DOX was loaded into the developed functionalized MWCNTs after proper characterization using dialysis diffusion method. The in vitro, ex vivo, and in vivo studies were performed on the MCF-7 cell line for assessment of the cancer targeting propensity. Both qualitative and quantitative cell uptake studies indicated the preferential higher uptake of estrone anchored nanotube formulation compared to other formulations and free DOX owing to the overexpression of estrogen receptors (ERs) on human breast MCF-7 cells. Similarly, the pharmacokinetic and increased antitumor activities also confirmed the elevated cancer targeting propensity of the estrone and folic acid anchored MWCNT formulations. The DOX/ES-PEG-MWCNTs has also shown significantly longer survival span (43 days) than free DOX (18 days) and control group (12 days). Present outcomes from the ex vivo and in vivo studies are deemed to be of great scientific value and shall assist targeted drug delivery formulation scientists for selection of the targeting moieties in the treatment of human breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Triethylamin, ≥99%
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Supelco
Aceton, analytical standard
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Aceton, histological grade, ≥99.5%