Merck
  • Comparison of PoraPak Rxn RP and XAD-2 adsorbents for monitoring dissolved hydrophobic organic contaminants.
Comparison of PoraPak Rxn RP and XAD-2 adsorbents for monitoring dissolved hydrophobic organic contaminants.
Environmental monitoring and assessment (2014-08-08)
Mark Omara, Thomas M Holsen, Xiaoyan Xia, James J Pagano, Bernard S Crimmins, Philip K Hopke
ZUSAMMENFASSUNG
Accurate determination of the levels of dissolved hydrophobic organic contaminants (HOCs) is an important step in estimating the dynamics of their inputs and losses in aqueous systems. This study explores an alternative method for efficiently sampling dissolved HOCs while mitigating a number of sampling artifacts associated with traditional methods. The adsorption characteristics of a new polymeric resin, PoraPak Rxn RP (PPR), were assessed using sorption isotherm experiments and fixed bed adsorption studies. The adsorption capacities and breakthrough times for four model contaminants (phenol, p-nitrophenol, naphthalene, and 2,4,6-tribromophenol) were proportional to the contaminant's hydrophobicity. The ability of PPR to isolate dissolved polychlorinated biphenyls (PCBs) in real samples was compared with that of XAD-2, a well-known macroporous polymer that suffers from high background contamination. The results indicated that the PPR resin can be effectively used for monitoring HOCs, with low ∑PCB levels in blanks, decreasing solvent use, and reducing extraction times.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
4-Nitrophenol -Lösung, 10 mM
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol -Lösung, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol -Lösung, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
2,4,6-Tribromphenol, 99%
Supelco
Naphthalin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)