Merck
  • X-linked inhibitor of apoptosis-associated factor l (XAFl) enhances the sensitivity of colorectal cancer cells to cisplatin.
X-linked inhibitor of apoptosis-associated factor l (XAFl) enhances the sensitivity of colorectal cancer cells to cisplatin.
Medical oncology (Northwood, London, England) (2014-11-05)
Wen-Cui Ju, Guo-Bin Huang, Xiao-Yong Luo, Wei-Hua Ren, De-Qing Zheng, Pin-Jia Chen, Yun-Feng Lou, Bin Li
ZUSAMMENFASSUNG
The purpose of present study was to investigate the roles of X-linked inhibitor of apoptosis-associated factor l (XAFl) in regulation apoptosis of colorectal cancer (CRC) cells after treatment with cisplatin (DDP). A total of ten paired cancerous and non-cancerous tissues were collected from patients with CRC after surgery. The levels of XAFl protein were detected by Western blot. Primary CRC cells were separated from cancer tissues, and its viability or apoptosis after treatment with DDP was determined with MTT or Annexin V/PI assays, respectively. Furthermore, we either up-regulated transfecting a XAF1 overexpression vector or down-regulated XAF1 by siRNA interference. And then, the XAF1 levels and its sensitivity to cisplatin were assessed. XAFl had a lower expression in the cancerous tissues from samples T1, T2 and T3 than their paired non-cancerous tissues N1, N2 and N3. However, the expression of XAF1 was not detected in samples T4 and N1. XAF1 levels in cancer tissues significantly decreased in comparison with normal tissues. Cell abilities of primary cells were significantly decreased in a dose-dependent manner, after treatment with a series concentrations of cisplatin (2, 5, 10 μg/mL) for 48 h. Although, after down-expression of XAFl by siRNA, cisplatin caused a significant decreases in apoptosis rates in CRC cells. The up-regulation of XAF1 distinctly increased apoptosis in CRC cells administered by cisplatin (P < 0.001). The XAFl could promoted apoptosis and enhanced chemotherapy sensitivity to cisplatin in CRC cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®