Merck
  • Amphotericin B transfer to CSF following intravenous administration of liposomal amphotericin B.
Amphotericin B transfer to CSF following intravenous administration of liposomal amphotericin B.
The Journal of antimicrobial chemotherapy (2014-06-04)
Volker Strenger, Andreas Meinitzer, Josef Donnerer, Nora Hofer, Hans Jürgen Dornbusch, Ulrike Wanz, Markus G Seidel, Daniela Sperl, Herwig Lackner, Wolfgang Schwinger, Petra Sovinz, Martin Benesch, Christian Urban
ZUSAMMENFASSUNG
Although amphotericin B (AmB) and its lipid formulations are used for the treatment of fungal infections of the CNS, the kinetics of AmB in the CSF after intravenous administration of liposomal amphotericin B (LAmB) are not well characterized. From 14 paediatric haemato-oncological patients (aged 0.4-19.5 years, median 7.6 years), we obtained 30 CSF samples by means of routine punctures (performed for intrathecal treatment of the underlying diseases) at different timepoints after the prophylactic intravenous infusion of LAmB (AmBisome, 3 mg/kg/day). Concurrent serum samples were obtained to calculate the transfer rates. An HPLC method was used for AmB detection. CSF levels of AmB 1-100 h after the intravenous infusion of LAmB were between 10 and 120 ng/mL, except in one case with a level of 529 ng/mL. Concurrent serum levels were about 1000-fold higher, ranging between 3 and 75 μg/mL. CSF levels did not show a clear time-dependent concentration profile, but remained at a steady-state for longer than 48 h after infusion. The transfer rate ranged from 0.02% to 0.92% (median 0.13%) and correlated significantly (r=0.801, P<0.001) with increasing time after infusion. After the intravenous administration of LAmB, AmB CSF levels were low, confirming published animal data. CSF levels remained at a steady-state level for longer than 48 h. As indicated by published post mortem data, higher levels in brain tissue, which would be necessary for the successful treatment of CNS infections, might be possible.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF