Merck
  • Structural characterization of secoiridoid glycosides by high-performance liquid chromatography/electrospray ionization mass spectrometry.
Structural characterization of secoiridoid glycosides by high-performance liquid chromatography/electrospray ionization mass spectrometry.
Rapid communications in mass spectrometry : RCM (2014-06-05)
Shufang Wang, Yimin Xu, Pinghong Chen, Yufeng Zhang
ZUSAMMENFASSUNG
The fragmentation behavior of seven secoiridoid glycosides (SIGs) isolated from Gentiana triflora Pall., a traditional Chinese medicine, was investigated by electrospray ionization ion trap mass spectrometry (ESI-IT-MS). Multistage electrospray ionization mass spectrometry (ESI-MS(n)) experiments both in positive- and negative-ion mode were used to elucidate the main fragmentation pathways of these compounds. In full scan mass spectra, different deprotonated or protonated molecules were observed for compounds with different basic structure. Ring cleavages of the aglycon moiety were observed in MS(2) spectra and the characteristic ions provided valuable information on the basic structural skeletons. Their fragmentation pathways and fragment ion structures were proposed. The established fragmentation patterns have been successfully used to identify eleven SIGs in the extract of Gentiana triflora Pall.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.