Merck
  • Carriers of the COMT Met/Met allele have higher degrees of hypnotizability, provided that they have good attentional control: a case of gene-trait interaction.
Carriers of the COMT Met/Met allele have higher degrees of hypnotizability, provided that they have good attentional control: a case of gene-trait interaction.
The International journal of clinical and experimental hypnosis (2014-08-03)
Christian Rominger, Elisabeth M Weiss, Simone Nagl, Harald Niederstätter, Walther Parson, Ilona Papousek
ZUSAMMENFASSUNG
Genetic factors may explain part of the interindividual variability in hypnotizability. A new avenue that may provide more comprehensive understanding of the phenotypic effects of genetic variations is the study of gene-trait interaction. In this study, the authors investigate the relationship of the dopamine-related COMT and the serotonin-related 5-HTTLPR polymorphisms to hypnotizability by taking individual differences in executive attention into account. Homozygosity for the COMT Met allele, putatively linked to the capability or proneness to dissociate from reality, was associated with high hypnotizability only if paired with high-attention ability. The finding can be integrated into hypnosis theory and represents a case of gene-trait interaction suggesting that investigating the effects of a gene in the context of relevant psychological traits may further elucidate gene-brain-behavior relationships.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®