Merck
  • Discovery of 2-[2-(5-nitrofuran-2-yl)vinyl]quinoline derivatives as a novel type of antimetastatic agents.
Discovery of 2-[2-(5-nitrofuran-2-yl)vinyl]quinoline derivatives as a novel type of antimetastatic agents.
Bioorganic & medicinal chemistry (2014-12-04)
Chih-Hua Tseng, Cherng-Chyi Tzeng, Chien-Chih Chiu, Chih-Yao Hsu, Chon-Kit Chou, Yeh-Long Chen
ZUSAMMENFASSUNG
A number of 2-furanylvinylquinoline derivatives were synthesized and evaluated for antiproliferative activities against the growth of four cancer cell lines including non-small cell lung cancer (A549 and H1299), breast cancer (MCF-7 and MDA-MB-231) and normal diploid embryonic lung cell line (MRC-5). Among them, (E)-6-methoxy-3-(4-methoxyphenyl)-2-[2-(5-nitrofuran-2-yl)vinyl]quinoline (10c) was found low cytotoxic in all cancer cells and normal cell. The aim of this study was to investigate the anti-invasive and anti-metastatic activity of compound 10c in H1299 human lung cancer cells. In this study, compound 10c inhibited the migration and invasion of cells in a concentration-dependent manner by wound healing assay and transwell invasion assay. Furthermore, the inhibition of both phosphorylation of Akt and ERK by compound 10c may be critical for cell migration and this may result in the down-regulation of several factors associated with cellular migration, including β-catenin transcription factor, Bcl-2 and COX-2. Interestingly, the treatment of compound 10c did not affect the expression level but reduced the activities of the MMP-2 and -9. The phosphorylation level of stress-activated kinase p38 was significantly increased following compound 10c treatment. To sum up, compound 10c had potential to suppress the migration and invasion of H1299 cancer cells in vitro and it could serve as a promising drug for the treatment of cancer metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®