Merck
  • MicroRNA-1269 promotes proliferation in human hepatocellular carcinoma via downregulation of FOXO1.
MicroRNA-1269 promotes proliferation in human hepatocellular carcinoma via downregulation of FOXO1.
BMC cancer (2014-12-05)
Xue-Wei Yang, Guan-Zhu Shen, Liang-Qi Cao, Xiao-Feng Jiang, He-Ping Peng, Gang Shen, De Chen, Ping Xue
ZUSAMMENFASSUNG
Hepatocellular carcinoma (HCC) is one of the most common malignancies and a major cause of cancer-related mortality in the world. MicroRNAs (miRNAs) are small, noncoding RNAs that play essential roles in various stages during cancer progression. The aim of the current study was to elucidate the role of miR-1269 in the pathogenesis of HCC. The expression of miR-1269 in HCC cells and tissues were determined by Real-time PCR analysis. Cell viability, colony formation and anchorage-independent growth ability assays were performed to examine cell proliferative capacity and tumorigenicity. Flow cytometry analysis was conducted to determine cell cycle progression. The expression of p21, CyclinD1, phosphorylated Rb, Rb and FOXO1 were examined by Western blotting analysis. Luciferase assay was used to determine whether FOXO1 is the direct target of miR-1269. miR-1269 was upregulated in HCC cells and tissues. Ectopic miR-1269 expression promoted, but inhibition of miR-1269 reduced, proliferation, tumorigenicity and cell cycle progression of HCC cells. Furthermore, we demonstrated that FOXO1 was a direct target of miR-1269. Suppression of FOXO1 by miR-1269 was associated with dysregulation of p21, cyclin D1, phosphorylated Rb and Ki67 expression, thereby playing an essential role in the growth of HCC cells. Our study indicated that overexpression of miR-1269 promotes cell proliferation in HCC through directly suppressing FOXO1, and functions as an oncomiR in HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®