Merck
Yin Yang 1 is a target of microRNA-34 family and contributes to gastric carcinogenesis.
Oncotarget (2014-06-28)
An-Ming Wang, Tzu-Ting Huang, Kai-Wen Hsu, Kuo-Hung Huang, Wen-Liang Fang, Muh-Hwa Yang, Su-Shun Lo, Chin-Wen Chi, Jing-Jer Lin, Tien-Shun Yeh
ZUSAMMENFASSUNG
Gastric cancer is the second leading cause of cancer-related death worldwide. Herein, we investigated the role of transcription factor Yin Yang 1 (YY1), a multi-functional protein, in tumorigenesis of gastric cancer cells. Results showed that YY1 contributed to gastric carcinogenesis of SC-M1 cells including growth, viability, and abilities of colony formation, migration, invasion, and tumorsphere formation. Levels of pluripotency genes CD44, Oct4, SOX-2, and Nanog were also up-regulated by YY1 in SC-M1 cells. Additionally, the 3'-untranslated region (3'-UTR) of YY1 mRNA was the target of microRNA-34 (miR-34) family consisting of miR-34a, miR-34b, and miR-34c. Overexpression of miR-34 family suppressed carcinogenesis through down-regulation of YY1 in NUGC-3 gastric cancer cells scarcely expressing miR-34 family. Alternatively, knockdown of miR-34 family promoted tumorigenesis via up-regulation of YY1 in SC-M1 and AZ521 gastric cancer cells with higher levels of miR-34 family. The miR-34 family also affected tumorsphere ultra-structure and inhibited the xenografted tumor growth as well as lung metastasis of SC-M1 cells through YY1. Expressions of miR-34a and miR-34c in gastric cancer tissues of patients were lower than those in normal tissues. Taken together, these results suggest that miR-34 family-YY1 axis plays an important role in the control of gastric carcinogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.