Merck
Alkylphenolic contaminants in the diet: Sparus aurata juveniles hepatic response.
General and comparative endocrinology (2014-07-01)
I Traversi, G Gioacchini, A Scorolli, D G Mita, O Carnevali, A Mandich
ZUSAMMENFASSUNG
A wide range of endocrine disrupter chemicals can mimic steroid hormones causing adverse health effects. Nonylphenol (NP) and t-octhylphenol (t-OP) are man-made alkylphenolic environmental contaminants possessing controversial endocrine disruption properties. This study has investigated the effects of NP and t-OP enriched diets on hepatic tissue and biotransformation activities in the liver. To this aim, sea bream juveniles were fed with commercial diet enriched with three different doses of NP (NP1: 5mg/kg bw, NP2: 50mg/kg bw and NP3: 100mg/kg bw) or t-OP (t-OP1: 5mg/kg bw, t-OP2: 50mg/kg bw and t-OP3: 100mg/kg bw) for 21 days. A significant increase of the hepatosomatic index was observed in NP1 and t-OP1. Alteration of liver morphology was observed in both NP and t-OP exposed juveniles although the most altered endpoints were observed in t-OP2 with 100% of tissue degeneration. Ethoxyresorufin-O-deethylase activity was significantly inhibited by NP and t-OP (p<0.05), while catalase activity was significantly induced, at both doses. A different pattern of protein expression of different isoforms of both vitellogenin and zona radiata protein was evidenced within the treatments. In addition, a significant increase in the abundance of the stress induced heat shock protein 70 gene in the liver of t-OP2 fish and a significant increase in the abundance of the estrogen induced cathepsin D gene in the liver of NP1 and t-OP2 fish, were observed. Finally, estradiol-17β (E2) and testosterone (T) plasma levels and E2/T showed significantly different patterns in NP and t-OP exposed against control fish.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Testosteron, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Testosteron, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Preise und Verfügbarkeit sind derzeit nicht verfügbar.