Merck
Evaluation of the intestinal permeability and cytotoxic effects of cylindrospermopsin.
Toxicon : official journal of the International Society on Toxinology (2014-09-03)
Diego A Fernández, M Carmen Louzao, Natalia Vilariño, Maria Fraga, Begoña Espiña, Mercedes R Vieytes, Luis M Botana
ZUSAMMENFASSUNG
Cylindrospermopsin is a freshwater and widespread cyanotoxin considered hazardous for human health. Climate change and eutrophication are the main factors influencing the increasing presence of cylindrospermopsin producers that can contaminate human and animal drinking waters, leading to a rise in ecological and human risk. In order to reach the bloodstream and thus the target receptor, an orally administered drug must first cross the intestinal barrier. The goal of this study was to examine the cylindrospermopsin intestinal permeability and its cellular effects on intestinal and hepatic cells. We explored the human intestinal permeability of cylindrospermopsin by performing in vitro permeation studies across the Caco-2 cell monolayer. Cell permeability data indicated a limited passage of the toxin through the intact intestinal epithelium in a time and concentration dependent way. Cylindrospermopsin induced neither damage on the integrity of the monolayer nor cytotoxicity in tests performed with Caco-2 even at micromolar concentration. Opposite, when hepatic Clone 9 cells were exposed to cylindrospermopsin, a noticeable cytotoxicity was observed being more marked at the higher concentrations used. In addition, this cell line showed alterations in reduced glutathione content due to cylindrospermopsin over time. Meanwhile glutamate cysteine ligase levels, the first rate-limiting enzyme of the glutathione route, showed a significant increase. Therefore our results indicate that cylindrospermopsin cytotoxicity is unrelated to protein inhibition or a decrease of reduced glutathione levels in Clone 9 cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumtetraborat Decahydrat, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Borsäure, ReagentPlus®, ≥99.5%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumtetraborat Decahydrat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Borsäure, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamin, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethanolamin, ≥99%
SAFC
L-Glutamin