Merck
  • Dependency of 2-methoxyestradiol-induced mitochondrial apoptosis on mitotic spindle network impairment and prometaphase arrest in human Jurkat T cells.
Dependency of 2-methoxyestradiol-induced mitochondrial apoptosis on mitotic spindle network impairment and prometaphase arrest in human Jurkat T cells.
Biochemical pharmacology (2015-03-04)
Seung Tae Lee, Ji Young Lee, Cho Rong Han, Yoon Hee Kim, Do Youn Jun, Dennis Taub, Young Ho Kim
ZUSAMMENFASSUNG
The present study sought to determine the correlation between 2-methoxyestradiol (2-MeO-E2)-induced cell cycle arrest and 2-MeO-E2-induced apoptosis. Exposure of Jurkat T cell clone (JT/Neo) to 2-MeO-E2 (0.5-1.0 μM) caused G2/M arrest, Bak activation, Δψm loss, caspase-9 and -3 activation, PARP cleavage, intracellular ROS accumulation, and apoptotic DNA fragmentation, whereas none of these events except for G2/M arrest were induced in Jurkat T cells overexpressing Bcl-2 (JT/Bcl-2). Under these conditions, Cdk1 phosphorylation at Thr-161 and dephosphorylation at Tyr-15, up-regulation of cyclin B1 expression, histone H1 phosphorylation, Cdc25C phosphorylation at Thr-48, Bcl-2 phosphorylation at Thr-56 and Ser-70, Mcl-1 phosphorylation at Ser-159/Thr-163, and Bim phosphorylation were detected irrespective of Bcl-2 overexpression. Concomitant treatment of JT/Neo cells with 2-MeO-E2 and the G1/S blocking agent aphidicolin resulted in G1/S arrest and abrogation of all apoptotic events, including Cdk1 activation, phosphorylation of Bcl-2, Mcl-1 and Bim, and ROS accumulation. The 2-MeO-E2-induced phosphorylation of Bcl-2 family proteins and mitochondrial apoptotic events were suppressed by a Cdk1 inhibitor, but not by an Aurora A kinase (AURKA), Aurora B kinase (AURKB), JNK, or p38 MAPK inhibitor. Immunofluorescence microscopic analysis revealed that 2-MeO-E2-induced mitotic arrest was caused by mitotic spindle network impairment and prometaphase arrest. Whereas 10-20 μM 2-MeO-E2 reduced the proportion of intracellular polymeric tubulin to monomeric tubulin, 0.5-5.0 μM 2-MeO-E2 increased it. These results demonstrate that the apoptogenic effect of 2-MeO-E2 (0.5-1.0 μM) was attributable to mitotic spindle defect-mediated prometaphase arrest, Cdk1 activation, phosphorylation of Bcl-2, Mcl-1, and Bim, and activation of Bak and mitochondria-dependent caspase cascade.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Adenosin-5′-diphosphoribose Natriumsalz, ≥93%
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Aphidicolin, analytical standard