Merck
  • Identification of the environmental neurotoxins annonaceous acetogenins in an Annona cherimolia Mill. Alcoholic Beverage Using HPLC-ESI-LTQ-Orbitrap.
Identification of the environmental neurotoxins annonaceous acetogenins in an Annona cherimolia Mill. Alcoholic Beverage Using HPLC-ESI-LTQ-Orbitrap.
Journal of agricultural and food chemistry (2014-08-05)
Jessica Le Ven, Isabelle Schmitz-Afonso, Guy Lewin, Alain Brunelle, David Touboul, Pierre Champy
ZUSAMMENFASSUNG
Epidemiological and toxicological studies have suggested Annonaceaeous acetogenins to be environmental neurotoxins responsible for sporadic atypical parkinsonism/dementia in tropical areas. These compounds are present in the tropical genus Annona (Annonaceae), known for its fruit-yielding cultivated species such as Annona cherimolia. This species is widely cultivated in South America, Spain, and Portugal and yields acetogenins in its seeds, stems, and roots. The presence of these compounds in the pulp of its fruit and in derived food products is unclear. An innovative and sensitive methodology by HPLC-ESI-LTQ-Orbitrap with postcolumn infusion of lithium iodide was used to identify the presence of low levels of acetogenins in an A. cherimolia Mill. fruit-based commercial alcoholic beverage. More than 80 representatives were detected, and the 31 most intense acetogenins were identified. All together these findings indicate that this species should be considered as a risk factor within the framework of a worldwide problem of food toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Lithiumiodid, 99.9% trace metals basis
Supelco
Methanol, analytical standard
Supelco
Ethylacetat, analytical standard