Merck
  • Tablet formulation of an active pharmaceutical ingredient with a sticking and filming problem: direct compression and dry granulation evaluations.
Tablet formulation of an active pharmaceutical ingredient with a sticking and filming problem: direct compression and dry granulation evaluations.
Drug development and industrial pharmacy (2013-11-28)
Naveen K Bejugam, Shravan K Mutyam, Gita N Shankar
ZUSAMMENFASSUNG
To develop a tablet formulation for an active pharmaceutical ingredient for which sticking and filming problems occurred during tablet punching. Direct compression and dry granulation tableting techniques were evaluated using factorial experimental design. The effects of chrome-coated punch tips, filler types and active percent in the tablet formulation by direct compression were evaluated. Similarly, for dry granulation using the roller compaction technique, three formulation factors - roller compaction pressure, intragranular filler percent and filler type - were studied. Tablets prepared by both techniques were characterized in regard to their compressibility index, tablet hardness, disintegration time, friability index and stickiness-filming index (an arbitrary index). Ten formulations were prepared by each technique. Using multiple response optimizations and estimated response surface plots, the data were analyzed to identify optimum levels for the formulation factors. Compressibility index values for all the formulations prepared by direct compression exceeded 25%, unlike the blends prepared by dry granulation. Both tablet hardness and disintegration time for direct compression formulations were significantly lower than for dry granulation formulations. The friability index values were significantly higher for direct compression formulations than for dry granulation formulations. All the direct compression formulations, unlike the dry granulation formulations, had a high stickiness-filming index. Statistical analysis helped in identifying the optimum levels of formulation factors, as well as the method for eliminating sticking and filming. Unlike the direct compression technique, dry granulation yielded tablets for which sticking and filming were completely eliminated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid