Merck
  • A nano-silver composite based on the ion-exchange response for the intelligent antibacterial applications.
A nano-silver composite based on the ion-exchange response for the intelligent antibacterial applications.
Materials science & engineering. C, Materials for biological applications (2014-06-09)
Chan Wang, Xiaobo Huang, Weilin Deng, Chengliang Chang, Ruiqiang Hang, Bin Tang
ZUSAMMENFASSUNG
As a kind of antimicrobial agent, nano-silver composites have attracted a great deal of interest in biomedical applications. However, the typical loadings of silver nanoparticles (AgNPs) in such composites could result in dose-related cytotoxicity. In this study, a nano-silver composite leading to antimicrobial activity without cytotoxicity was fabricated by loading AgNPs into a dried alginate hydrogel. The biological performance of this composite mainly depended on the release of AgNPs, which needed to be triggered by the ion-exchange response and was further influenced by the loadings of AgNPs in the composite. The antimicrobial activity against E. coli and S. aureus demonstrated that the released silver no less than 678 ppb in the medium caused a reduction of 7log10CFU/mL (100%) bacteria. Significantly, the dose (~1.10×10(3) ppb) of released silver was not toxic and allowed attachment, and growth of MC3T3-E1 pre-osteoblast cells. These results supported that the composite was compatible with in vitro mammalian cells yet exhibited antimicrobial activity by carefully designing the loadings of AgNPs within the alginate. Thus, it indicated that the performance of this composite might permit management of bacterial infection in wound beds without impairment of wound healing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®