Merck
Photochemical degradation of Corexit components in ocean water.
Chemosphere (2014-07-07)
Caitlin M Glover, Stephen P Mezyk, Karl G Linden, Fernando L Rosario-Ortiz
ZUSAMMENFASSUNG
Due to the large quantities of dispersants used during the Deepwater Horizon spill in 2010, there were immediate concerns with regards to the fate and transport of the mixture in ocean waters. Direct and sensitized photolysis experiments were carried out for two compounds chosen as surrogates for the Corexit mixture (9500 and 9527) that were applied to surface waters during the oil spill in the Gulf of Mexico. The results showed that direct photolysis did not contribute significantly to the overall degradation (max ∼30%), therefore the focus shifted to sensitized photolysis, specifically the degradation stemming from the reaction rate with hydroxyl radical (HO). The direct photochemical degradation rates for two of the compounds, dioctyl sulfosuccinate (DOSS) and dipropylene glycol butyl ether (DGBE) were measured as 4.29×10(-6)s(-1) and 5.95×10(-6)s(-1), respectively; whereas the overall degradation rate in ocean water was 1.56×10(-5)s(-1) and 2.23×10(-5)s(-1). The formation rates and apparent quantum yields for HO formation were determined for six ocean water samples. The values ranged from 1.81×10(-5) near shore to 0.061×10(-5) for the open ocean. These degradation rates suggest the possibility for photolysis to play a role in the overall fate of Corexit.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Methanol, analytical standard
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%