Merck
  • Enhancement of enzymatic colorimetric response by silver island films on high throughput screening microplates.
Enhancement of enzymatic colorimetric response by silver island films on high throughput screening microplates.
Journal of immunological methods (2014-06-21)
Biebele Abel, Travis C Clement, Kadir Aslan
ZUSAMMENFASSUNG
In this study, we report the use of an enzyme-based hybrid platform, which is comprised of silver island films, enzymes (HRP and AP) and high-throughput screening (HTS) microplates, to enhance the colorimetric response of enzymatic reactions. The hybrid platform was designed in a two-step process: (i) deposition of SIFs onto HTS microplates with low, medium, and high loading (refers to the extent of the surface plasmon resonance peak of SIFs at 460 nm) using Tollen's reaction scheme; and (ii) attachment of b-BSA or BEA as linkers for the immobilization of enzymes. The presence of SIFs within the wells of the HTS microplates was confirmed using an optical spectrophotometer and real-color photography. Control experiments, where SIFs were omitted from the surfaces were carried out to confirm the effect of SIFs on the enzymatic colorimetric response. Significant colorimetric signal enhancement was observed for HRP or AP on SIFs (high loading) deposited HTS microplates using b-BSA (up to ~3-fold for AP and ~6-fold HRP) or BEA (up to ~7-fold for both HRP and AP), as compared to our control samples. The observed increase in colorimetric response can be attributed to the nature of BEA, which exposes surface-bound enzymes to the substrate present in bulk more efficiently than b-BSA. This study proves that SIFs can serve as a valuable tool to improve the signal output of existing bioassays carried out in HTS microplates, which can be applicable to the field biosensors and plasmonics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Natriumphosphat, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Borsäure, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumphosphat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Borsäure, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%