Merck
Toxicokinetics of permethrin biomarkers of exposure in orally exposed volunteers.
Toxicology letters (2014-12-17)
Mylène Ratelle, Jonathan Côté, Michèle Bouchard
ZUSAMMENFASSUNG
Permethrin is a widely used pyrethroid insecticide for which the toxicokinetics of exposure biomarkers in humans is not fully documented. The time courses of key biomarkers of permethrin exposure were thus assessed in accessible biological matrices of orally exposed volunteers. Six volunteers ingested 0.1 mg/kg body weight of permethrin (60:40 trans/cis). Blood samples were withdrawn at fixed periods over 72 h following ingestion and complete timed-urine voids were collected over 84 h post-dosing. Cis-and trans-3-(2,2-dichlorovinyl)-2,2-dimethyl-cyclopropane-1-carboxylic acids (cis-and trans-DCCA) and 3-phenoxybenzoic acid (3-PBA) were quantified in samples. In plasma, peak concentrations of cis-DCCA, trans-DCCA and 3-PBA were reached about ≈7 h post-dosing, and elimination appeared monophasic with a mean apparent elimination half-life (t½) of 6.2, 7.1 and 6.5 h, respectively. In urine, elimination rate time courses of cis-DCCA, trans-DCCA and 3-PBA evolved in parallel with plasma, with respective mean apparent elimination t½ of 4.5, 5.4 and 5.7 h. Over the 84 h period post-treatment, 43-46% of administered molar dose were excreted in urine as trans-DCCA (molar % of trans-permethrin) and 3-PBA. Results show similarities in the different metabolite profiles and a rapid equilibrium between urine and plasma levels; data should help interpret the significance of biological measurements and optimal sampling strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Ethylacetat, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard