Merck
  • Inhibition of CD39 enzymatic function at the surface of tumor cells alleviates their immunosuppressive activity.
Inhibition of CD39 enzymatic function at the surface of tumor cells alleviates their immunosuppressive activity.
Cancer immunology research (2014-11-19)
Jeremy Bastid, Anne Regairaz, Nathalie Bonnefoy, Cécile Déjou, Jérôme Giustiniani, Caroline Laheurte, Stéphanie Cochaud, Emilie Laprevotte, Elisa Funck-Brentano, Patrice Hemon, Laurent Gros, Nicole Bec, Christian Larroque, Gilles Alberici, Armand Bensussan, Jean-François Eliaou
ZUSAMMENFASSUNG
The ectonucleotidases CD39 and CD73 hydrolyze extracellular adenosine triphosphate (ATP) and adenosine diphosphate (ADP) to generate adenosine, which binds to adenosine receptors and inhibits T-cell and natural killer (NK)-cell responses, thereby suppressing the immune system. The generation of adenosine via the CD39/CD73 pathway is recognized as a major mechanism of regulatory T cell (Treg) immunosuppressive function. The number of CD39⁺ Tregs is increased in some human cancers, and the importance of CD39⁺ Tregs in promoting tumor growth and metastasis has been demonstrated using several in vivo models. Here, we addressed whether CD39 is expressed by tumor cells and whether CD39⁺ tumor cells mediate immunosuppression via the adenosine pathway. Immunohistochemical staining of normal and tumor tissues revealed that CD39 expression is significantly higher in several types of human cancer than in normal tissues. In cancer specimens, CD39 is expressed by infiltrating lymphocytes, the tumor stroma, and tumor cells. Furthermore, the expression of CD39 at the cell surface of tumor cells was directly demonstrated via flow cytometry of human cancer cell lines. CD39 in cancer cells displays ATPase activity and, together with CD73, generates adenosine. CD39⁺CD73⁺ cancer cells inhibited the proliferation of CD4 and CD8 T cells and the generation of cytotoxic effector CD8 T cells (CTL) in a CD39- and adenosine-dependent manner. Treatment with a CD39 inhibitor or blocking antibody alleviated the tumor-induced inhibition of CD4 and CD8 T-cell proliferation and increased CTL- and NK cell-mediated cytotoxicity. In conclusion, interfering with the CD39-adenosine pathway may represent a novel immunotherapeutic strategy for inhibiting tumor cell-mediated immunosuppression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Guanosin, ≥98%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Natriumchromat, 98%
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanosin, BioReagent, suitable for cell culture
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Guanosin, ≥97.0% (HPLC)
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Anti-ENTPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution