Merck
  • Fabrication of extended-release patient-tailored prednisolone tablets via fused deposition modelling (FDM) 3D printing.
Fabrication of extended-release patient-tailored prednisolone tablets via fused deposition modelling (FDM) 3D printing.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-12-03)
Justyna Skowyra, Katarzyna Pietrzak, Mohamed A Alhnan
ZUSAMMENFASSUNG
Rapid and reliable tailoring of the dose of controlled release tablets to suit an individual patient is a major challenge for personalized medicine. The aim of this work was to investigate the feasibility of using a fused deposition modelling (FDM) based 3D printer to fabricate extended release tablet using prednisolone loaded poly(vinyl alcohol) (PVA) filaments and to control its dose. Prednisolone was loaded into a PVA-based (1.75 mm) filament at approximately 1.9% w/w via incubation in a saturated methanolic solution of prednisolone. The physical form of the drug was assessed using differential scanning calorimetry (DSC) and X-ray powder diffraction (XRPD). Dose accuracy and in vitro drug release patterns were assessed using HPLC and pH change flow-through dissolution test. Prednisolone loaded PVA filament demonstrated an ability to be fabricated into regular ellipse-shaped solid tablets using the FDM-based 3D printer. It was possible to control the mass of printed tablet through manipulating the volume of the design (R(2) = 0.9983). On printing tablets with target drug contents of 2, 3, 4, 5, 7.5 and 10mg, a good correlation between target and achieved dose was obtained (R(2) = 0.9904) with a dose accuracy range of 88.7-107%. Thermal analysis and XRPD indicated that the majority of prednisolone existed in amorphous form within the tablets. In vitro drug release from 3D printed tablets was extended up to 24h. FDM based 3D printing is a promising method to produce and control the dose of extended release tablets, providing a highly adjustable, affordable, minimally sized, digitally controlled platform for producing patient-tailored medicines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Prednisolon, ≥99%