Merck
  • Aberrant neuronal differentiation and inhibition of dendrite outgrowth resulting from endoplasmic reticulum stress.
Aberrant neuronal differentiation and inhibition of dendrite outgrowth resulting from endoplasmic reticulum stress.
Journal of neuroscience research (2014-04-12)
Koichi Kawada, Takaaki Iekumo, Ryo Saito, Masayuki Kaneko, Seisuke Mimori, Yasuyuki Nomura, Yasunobu Okuma
ZUSAMMENFASSUNG
Neural stem cells (NSCs) play an essential role in development of the central nervous system. Endoplasmic reticulum (ER) stress induces neuronal death. After neuronal death, neurogenesis is generally enhanced to repair the damaged regions. However, it is unclear whether ER stress directly affects neurogenesis-related processes such as neuronal differentiation and dendrite outgrowth. We evaluated whether neuronal differentiation and dendrite outgrowth were regulated by HRD1, a ubiquitin ligase that was induced under mild conditions of tunicamycin-induced ER stress. Neurons were differentiated from mouse embryonic carcinoma P19 cells by using retinoic acid. The differentiated cells were cultured for 8 days with or without tunicamycin and HRD1 knockdown. The ER stressor led to markedly increased levels of ER stress. ER stress increased the expression levels of neuronal marker βIII-tubulin in 8-day-differentiated cells. However, the neurites of dendrite marker microtubule-associated protein-2 (MAP-2)-positive cells appeared to retract in response to ER stress. Moreover, ER stress markedly reduced the dendrite length and MAP-2 expression levels, whereas it did not affect the number of surviving mature neurons. In contrast, HRD1 knockdown abolished the changes in expression of proteins such as βIII-tubulin and MAP-2. These results suggested that ER stress caused aberrant neuronal differentiation from NSCs followed by the inhibition of neurite outgrowth. These events may be mediated by increased HRD1 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥99% (GC), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Retinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Desoxy-D-Glucose, ≥98% (GC), BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.