Merck
  • Modulatory effects of extracts of vinegar-baked Radix Bupleuri and saikosaponins on the activity of cytochrome P450 enzymes in vitro.
Modulatory effects of extracts of vinegar-baked Radix Bupleuri and saikosaponins on the activity of cytochrome P450 enzymes in vitro.
Xenobiotica; the fate of foreign compounds in biological systems (2014-05-02)
Tongya Yu, Xianzhi Chen, Yinjie Wang, Ruizhi Zhao, Shirui Mao
ZUSAMMENFASSUNG
1. In this article, the modulatory effects of extracts from vinegar-baked Radix Bupleuri (VBRB) and saikosaponins on the activity of CYP1A2, CYP2C9 and CYP3A4 were investigated in vitro. 2. Microsomal in vitro incubation method was utilized to simulate metabolic reaction under physiological environment by incubating the marker with liver microsomes in the absence or presence of VBRB and saikosaponins. The contents of 4-acetamidophenol, 6β-hydroxyltestosterone and 4-hydroxydiclofenac, the metabolites of phenacetin, testosterone and diclofenac, which were selected as specific probe drugs of CYP1A2, CYP2C9 and CYP3A4, respectively, were analyzed by high-performance liquid chromatography. 3. The production of the metabolites was incubation time dependent. The modulatory effects of different VBRB extracts and saikosaponins on CYP isoforms increased with concentration. Among all the extracts studied, BC1 has a strong inhibition effect compared to the three CYP isoforms tested, while the others have only significant inhibition on the activity of CYP2C9. 4. This in vitro study demonstrated that various extracts of VBRB tested in this study have negligible potential to interfere with CYP1A2- and CYP3A4-metabolized drugs; risk of herb-drug interaction might occur when VBRB is concurrently taken with CYP2C9 substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4-Acetamidophenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-Acetamidophenol, BioXtra, ≥99.0%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Hydrat
Sigma-Aldrich
Diclofenac Natriumsalz
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Sigma-Aldrich
4-Acetamidophenol, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Diclofenac Natriumsalz, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Acetamidophenol, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenacetin Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Diclofenacnatrium, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Diclofenac Natriumsalz, analytical standard
USP
Phenacetin-Schmelzpunktstandard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 5 mg (per vial)
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Diclofenac für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Diclofenac-Natrium, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosteron, VETRANAL®, analytical standard
Testosteron, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 10 mg (per vial)