Merck
  • Requirement for autophagy in the long-term persistence but not initial formation of memory B cells.
Requirement for autophagy in the long-term persistence but not initial formation of memory B cells.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-13)
Min Chen, Srikanth Kodali, Albert Jang, Le Kuai, Jin Wang
ZUSAMMENFASSUNG
Autophagy is required for the long-term maintenance of Ag-specific memory B cells. However, whether autophagy is also important for the initial formation of memory B cells remains unclear. In this study, we show that newly generated memory B cells do not display active autophagy but are capable of forming Ab-secreting cells after rechallenge with Ags. Increases in autophagy took place over time after the initial formation of memory B cells. The expression of transcription factors involved in autophagy, but not changes in epigenetic regulation by DNA methylation, was required for autophagy gene expression and the development of active autophagy in memory B cells. This indicates that autophagy is not critical for the initial generation of memory B cells but is required for their long-term persistence. Our results suggest that promoting autophagy to improve Ab-dependent immunological memory is more effective during memory B cell maintenance stage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, ACS reagent, ≥98%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Wasser, BioPerformance Certified
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Wasser, endotoxin, free
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioXtra, ≥98.0%
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O