Merck
  • Gaucher disease due to saposin C deficiency is an inherited lysosomal disease caused by rapidly degraded mutant proteins.
Gaucher disease due to saposin C deficiency is an inherited lysosomal disease caused by rapidly degraded mutant proteins.
Human molecular genetics (2014-06-14)
Marialetizia Motta, Serena Camerini, Massimo Tatti, Marialuisa Casella, Paola Torreri, Marco Crescenzi, Marco Tartaglia, Rosa Salvioli
ZUSAMMENFASSUNG
Saposin (Sap) C is an essential cofactor for the lysosomal degradation of glucosylceramide (GC) by glucosylceramidase (GCase) and its functional impairment underlies a rare variant form of Gaucher disease (GD). Sap C promotes rearrangement of lipid organization in lysosomal membranes favoring substrate accessibility to GCase. It is characterized by six invariantly conserved cysteine residues involved in three intramolecular disulfide bonds, which make the protein remarkably stable to acid environment and degradation. Five different mutations (i.e. p.C315S, p.342_348FDKMCSKdel, p.L349P, p.C382G and p.C382F) have been identified to underlie Sap C deficiency. The molecular mechanism by which these mutations affect Sap C function, however, has not been delineated in detail. Here, we characterized biochemically and functionally four of these gene lesions. We show that all Sap C mutants are efficiently produced, and exhibit lipid-binding properties, modulatory behavior on GCase activity and subcellular localization comparable with those of the wild-type protein. We then delineated the structural rearrangement of these mutants, documenting that most proteins assume diverse aberrant disulfide bridge arrangements, which result in a substantial diminished half-life, and rapid degradation via autophagy. These findings further document the paramount importance of disulfide bridges in the stability of Sap C and provide evidence that accelerated degradation of the Sap C mutants is the underlying pathogenetic mechanism of Sap C deficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2,5-Dihydroxybenzoesäure, 98%
Supelco
2,5-Dihydroxybenzoesäure, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Supelco
2,5-Dihydroxybenzoesäure, analytical standard
Supelco
2,5-Dihydroxybenzoesäure, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material