Merck
Induction of Herpud1 expression by ER stress is regulated by Nrf1.
FEBS letters (2015-02-01)
Daniel V Ho, Jefferson Y Chan
ZUSAMMENFASSUNG
Herpud1 is an ER-localized protein that contributes to endoplasmic reticulum (ER) homeostasis by participating in the ER-associated protein degradation pathway. The Nrf1 transcription factor is important in cellular stress pathways. We show that loss of Nrf1 function results in decreased Herpud1 expression in cells and liver tissues. Expression of Herpud1 increases in response to ER stress, but not in Nrf1 knockout cells. Transactivation studies show that Nrf1 acts through antioxidant response elements located in the Herpud1 promoter, and chromatin immunoprecipitation demonstrates that Herpud1 is a direct Nrf1 target gene. These results indicate that Nrf1 is a transcriptional activator of Herpud1 expression during ER stress, and they suggest Nrf1 is a key player in the regulation of the ER stress response in cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Menadion, crystalline
Sigma-Aldrich
Menadion, meets USP testing specifications
Supelco
Menadion (K3), analytical standard
Supelco
Menadion, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Menadion, United States Pharmacopeia (USP) Reference Standard
Menadion, European Pharmacopoeia (EP) Reference Standard