Merck
Structural and functional assessment of perilipin 2 lipid binding domain(s).
Biochemistry (2014-10-23)
Charles P Najt, Joel S Lwande, Avery L McIntosh, Subramanian Senthivinayagam, Shipra Gupta, Leslie A Kuhn, Barbara P Atshaves
ZUSAMMENFASSUNG
Although perilipin 2 (Plin2) has been shown to bind lipids with high affinity, the Plin2 lipid binding site has yet to be defined. This is of interest since Plin2's affinity for lipids has been suggested to be important for lipid droplet biogenesis and intracellular triacylglycerol accumulation. To define these regions, mouse Plin2 and several deletion mutants expressed as recombinant proteins and in mammalian cells were assessed by molecular modeling, fluorescence binding, circular dichroic, and fluorescence resonance energy transfer techniques to identify the structural and functional requirements for lipid binding. Major findings of this study indicate (1) the N-terminal PAT domain does not bind cholesterol or stearic acid; (2) Plin2 residues 119-251, containing helix α4, the α-β domain, and part of helix α6 form a Plin3-like cleft found to be important for highest affinity lipid binding; (3) both stearic acid and cholesterol interact favorably with the Plin2 cleft formed by conserved residues in helix α6 and adjacent strands, which is common to all the active lipid-binding constructs; and (4) discrete targeting of the Plin2 mutants to lipid droplets supports Plin2 containing two independent, nonoverlapping lipid droplet targeting domains in its central and C-terminal sequences. Thus, the current work reveals specific domains responsible for Plin2-lipid interactions that involves the protein's lipid binding and targeting functions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
USP
Stearinsäure, United States Pharmacopeia (USP) Reference Standard
Supelco
Stearinsäure, analytical standard
Supelco
Stearinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Stearinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.