Merck
  • Sub-inhibitory concentrations of antifungals suppress hemolysin activity of oral Candida albicans and Candida tropicalis isolates from HIV-infected individuals.
Sub-inhibitory concentrations of antifungals suppress hemolysin activity of oral Candida albicans and Candida tropicalis isolates from HIV-infected individuals.
Mycopathologia (2014-08-22)
Sukumaran Anil, Mohamed Hashem, Sajith Vellappally, Shankargouda Patil, H M H N Bandara, L P Samaranayake
ZUSAMMENFASSUNG
Secretion of hydrolytic enzymes such as hemolysin is considered an important virulence attribute of the opportunistic pathogenic fungus Candida. It is known that Candida spp. isolated from HIV-infected patients produce copious hemolysins. As common antifungal agents may perturb the production of extracellular enzymes, we evaluated the effect of three antifungals nystatin, amphotericin B and fluconazole on the hemolytic activity of Candida albicans and Candida tropicalis isolates from HIV-infected individuals. The impact of antimycotics on hemolytic activity was assessed by a previously described in vitro plate assay, after exposing ten isolates each of C. albicans and C. tropicalis recovered from HIV-infected individuals to sub-minimum inhibitory concentrations (sub-MIC) of nystatin, amphotericin B and fluconazole. All Candida isolates showed a significant reduction in hemolytic activity. The reduction was highest for amphotericin B-exposed C. albicans and C. tropicalis followed by nystatin and fluconazole. The effect of antimycotics was more pronounced on the hemolytic activity of C. tropicalis compared to that of C. albicans. Commonly used antifungal agents significantly suppress hemolysin activity of Candida species. This implies that the antifungals, in addition to their lethality, may modulate key virulence attributes of the yeast. The clinical relevance of this phenomenon in HIV disease and other similar pathologies remains to be determined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Fluconazol, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Stickstoff, ≥99.998%
Supelco
Fluconazol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
Supelco
Dimethylsulfoxid, analytical standard
USP
Fluconazol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Fluconazol, European Pharmacopoeia (EP) Reference Standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Fluconazol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®