Merck
Berberine inhibits the proliferation of human uterine leiomyoma cells.
Fertility and sterility (2015-02-17)
Hsiao-Li Wu, Tung-Yueh Chuang, Ayman Al-Hendy, Michael P Diamond, Ricardo Azziz, Yen-Hao Chen
ZUSAMMENFASSUNG
To determine whether berberine (BBR), a naturally occurring plant-derived alkaloid, inhibits the proliferation of human uterine leiomyoma (UtLM) cells. Laboratory research. Laboratory. UtLM and normal human uterine smooth muscle (UtSMC) cell lines. Treatment with [1] BBR (10, 20, and 50 μM), [2] BBR (20 and 50 μM) and/or 17β-estradiol (E2; 10 and 100 nM), and [3] BBR (20 and 50 μM) and/or progesterone (P4; 10 and 100 nM) for 24 or 72 hours. Cell proliferation, cell cycle, apoptosis, and related genes expression were determined. BBR inhibited UtLM cell proliferation by inducing G2/M cell cycle arrest and apoptosis. Cell cycle G2/M phase-related genes were altered by BBR treatment: the expression of cyclin A1, cyclin B1, and Cdk1 were down-regulated, while Cdk4, p21, and p53 were up-regulated. BBR-treated cells stained positively for annexin V and manifested increased BAX expression. E2- and P4-induced UtLM cell proliferation was blocked by BBR treatment. In marked contrast, even the highest concentration of BBR (50 μM) did not influence cell proliferation in UtSMC cells. BBR selectively inhibits cellular proliferation and blocks E2- and P4-induced cell proliferation in UtLM but not in normal UtSMC cells. In addition, BBR did not demonstrate cytotoxicity effects in normal human UtSMCs. Our results suggest BBR could be a potential therapeutic agent for the treatment of uterine leiomyoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Berberin Chlorid-Form
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture