Merck
  • Expectorant, antitussive, anti-inflammatory activities and compositional analysis of Aster tataricus.
Expectorant, antitussive, anti-inflammatory activities and compositional analysis of Aster tataricus.
Journal of ethnopharmacology (2015-02-24)
Ping Yu, Si Cheng, Juan Xiang, Bin Yu, Mian Zhang, Chaofeng Zhang, Xianghong Xu
ZUSAMMENFASSUNG
The root of Aster tataricus L. f., recorded in all versions of Chinese Pharmacopoeia, is a traditional Chinese medicine with the function of dispelling phlegm and relieving cough for more than 2000 years. This study was designed to evaluate the expectorant, antitussive, and anti-inflammatory activities of the root of A. tataricus and to explore the chemical substances responsible for these activities. The 70% ethanol extract of the root of A. tataricus (RA-70) was divided into three fractions, Fr-0, Fr-50 and Fr-95. They were all orally administrated to the mice to investigate their potential expectorant activities by a tracheal phenol red secretion method. The most effective fraction, together with shionone, was evaluated the expectorant, antitussive and anti-inflammatory activities by the mouse models of phenol red secretion, ammonia-induced cough, and xylene-induced ear swelling. Furthermore, the chemical components of the effective fraction were analyzed and identified by an HPLC-Q-TOF/MS method. Treatment with RA-70, Fr-0 and Fr-50 increased the amount of phenol red secretion by 65.3%, 56.5%, and 76.9%, respectively. Fr-50 was chosen for the further investigation and the results showed that Fr-50 at 40, 80 mg/kg significantly enhanced the phenol red secretion of tracheas, increased the latent period and decreased the frequency of cough and inhibited the ear edema in mice. Shionone at 80 mg/kg showed the trend of enhancing sputum secreting, but had no effect on ammonia-induced cough and xylene-induced ear edema. HPLC-Q-TOF/MS analysis indicated that Fr-50 was mainly composed of 12 caffeoylquinic acids (40.8%, in relative peak area), 7 astersaponins (12.0%) and 13 astins/asterinins (pentapeptides, 26.5%). The root of A. tataricus has significant expectorant, antitussive and anti-inflammatory effects. Caffeoylquinic acids, astersaponins, and aster peptides, rather than shionone, may be the main constituents responsible for the expectorant and antitussive activities of A. tataricus and act in a synergistic way.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chlorogensäure, ≥95% (titration)
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material