Merck
  • Pharmacokinetics of tolbutamide and its metabolite 4-hydroxy tolbutamide in poloxamer 407-induced hyperlipidemic rats.
Pharmacokinetics of tolbutamide and its metabolite 4-hydroxy tolbutamide in poloxamer 407-induced hyperlipidemic rats.
Biopharmaceutics & drug disposition (2014-03-05)
Mi Ran Choi, Mi Hye Kwon, Yong Yeon Cho, Hye Duck Choi, Yu Chul Kim, Hee Eun Kang
ZUSAMMENFASSUNG
Under hyperlipidemic conditions, there are likely to be alterations in the pharmacokinetics of CYP2C11 substrates following decreased expression of CYP2C11, which is homologous to human CYP2C9. The pharmacokinetics of tolbutamide (TB) and its metabolite 4-hydroxy tolbutamide (4-OHTB) were evaluated as a CYP2C11 probe after intravenous and oral administration of 10 mg/kg tolbutamide to poloxamer 407-induced hyperlipidemic rats (HL rats). Changes in the expression and metabolic activity of hepatic CYP2C11 and the plasma protein binding of tolbutamide in HL rats were also evaluated. The total area under the plasma concentration-time curve (AUC) of tolbutamide in HL rats after intravenous administration was comparable to that in controls due to their comparable non-renal clearance (CLNR ). The free fractions of tolbutamide in plasma were comparable between the control and HL rats. The 4-hydroxylated metabolite formation ratio (AUC4-OHTB /AUCTB ) in HL rats was significantly smaller than that in the control rats as a result of the reduced expression of hepatic CYP2C11 (by 15.0%) and decreased hepatic CLint (by 28.8%) for metabolism of tolbutamide to 4-OHTB via CYP2C11. Similar pharmacokinetic changes were observed in HL rats after oral administration of tolbutamide. These findings have potential therapeutic implications, assuming that the HL rat model qualitatively reflects similar changes in patients with hyperlipidemia. Since other sulfonylureas in clinical use are substrates of CYP2C9, their hepatic CLint changes have the potential to cause clinically relevant pharmacokinetic changes in a hyperlipidemic state.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Supelco
Tromethamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Trizma® Base, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Tolbutamid, analytical standard
USP
Tromethamin, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard