Merck
  • Combined effects of estrogenic chemicals with the same mode of action using an estrogen receptor binding bioassay.
Combined effects of estrogenic chemicals with the same mode of action using an estrogen receptor binding bioassay.
Environmental toxicology and pharmacology (2014-12-03)
Rong Yang, Na Li, Mei Ma, Zijian Wang
ZUSAMMENFASSUNG
The increasing amounts of various estrogenic chemicals coexisting in the aquatic environment may pose environmental risks. While the concept of estradiol equivalent (EEQ) has been frequently applied in studying estrogenic mixtures, few experiments have been done to prove its reliability. In this study, the reliability of EEQ and the related model concentration addition (CA) was verified based on the two-hybrid recombinant yeast bioassay when all mixture components had the same mode of action and target of action. Our results showed that the measured estrogenic effects could be well predicted by CA and EEQ for all laboratory-made mixtures using two designs, despite the varying estrogenic activity, concentration levels and ratios of the test chemicals. This suggests that when an appropriate endpoint and its relevant bioassay are chosen, CA should be valid and the application of EEQ in predicting the effect of non-equi-effect mixtures is feasible.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
4,4′-Isopropylidendiphenol, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
4,4′-Isopropylidendiphenol, 97%
Sigma-Aldrich
Estron, ≥99%
Sigma-Aldrich
4-tert-Octylphenol, 97%
Supelco
4,4′-Isopropylidendiphenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Estron, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Estriol, ≥97%
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Estradiol 17-Valerat, ≥98%
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Supelco
4-tert-Octylphenol, analytical standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
USP
Estron, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estriol, meets USP testing specifications
Estradiolvalerat, European Pharmacopoeia (EP) Reference Standard