Merck
Activation of autophagy in response to nanosecond pulsed electric field exposure.
Biochemical and biophysical research communications (2015-02-11)
Jody C Ullery, Melissa Tarango, Caleb C Roth, Bennett L Ibey
ZUSAMMENFASSUNG
Previous work demonstrated significant changes in cellular membranes following exposure of cells to nanosecond pulsed electric fields (nsPEF), including nanoporation and increases in intracellular calcium concentration. While it is known that nsPEF exposure can cause cell death, how cells repair and survive nsPEF-induced cellular damage is not well understood. In this paper, we investigated whether autophagy is stimulated following nsPEF exposure to repair damaged membranes, proteins, and/or organelles in a pro-survival response. We hypothesized that autophagy is activated to repair nsPEF-induced plasma membrane damage and overwhelming this compensatory mechanism results in cell death. Activation of autophagy and subsequent cell death pathways were assessed measuring toxicity, gene and protein expression of autophagy markers, and by monitoring autophagosome formation and maturation using fluorescent microscopy. Results show that autophagy is activated at subtoxic nsPEF doses, as a compensatory mechanism to repair membrane damage. However, prolonged exposure results in increased cell death and a concomitant decrease in autophagic markers. These results suggest that cells take an active role in membrane repair, through autophagy, following exposure to nsPEF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles