Merck
  • Simultaneous determination of major type B trichothecenes and deoxynivalenol-3-glucoside in animal feed and raw materials using improved DSPE combined with LC-MS/MS.
Simultaneous determination of major type B trichothecenes and deoxynivalenol-3-glucoside in animal feed and raw materials using improved DSPE combined with LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-18)
Zhiyong Zhao, Qinxiong Rao, Suquan Song, Na Liu, Zheng Han, Jiafa Hou, Aibo Wu
ZUSAMMENFASSUNG
A simple and reliable method for simultaneous determination of deoxynivalenol-3-glucoside and major type B trichothecenes (deoxynivalenol, nivalenol, fusarenon X, 3-acetyldeoxynivalenol, 15-acetyldeoxynivalenol and deepoxy-deoxynivalenol) in animal feed and raw materials has been developed and validated in this study. The method was based on an improved dispersive solid-phase extraction (DSPE) followed by analysis using high performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS). Also, matrix-matched calibration curve (R(2)>0.99) was employed to minimize matrix effects and ensure accurate quantification. The recoveries during sample preparation process (including extraction and clean-up) ranged from 79.03% to 118.39%, with intra-day and inter-day relative standard deviation lower than 20% for all the analytes. The limit of quantification ranged from 5.0 μg/kg for deoxynivalenol to 13.6 μg/kg for fusarenon X. The validated method was successfully applied to the analysis of animal feed and corn. The pilot study showed that 37 out of 41 samples were contaminated with deoxynivalenol-3-glucoside at the levels of 6.0-121.0 μg/kg. Most of the type B trichothecenes were also found with the exception of fusarenon X, at the contaminated levels of 10.0-1,382 μg/kg. To the best of our knowledge, this was the first scientific report on the co-occurrence of masked deoxynivalenol and type B trichothecenes in animal feed and raw materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.