Merck
  • Cutaneous drug delivery of capsaicin after in vitro administration of the 8% capsaicin dermal patch system.
Cutaneous drug delivery of capsaicin after in vitro administration of the 8% capsaicin dermal patch system.
Skin pharmacology and physiology (2014-10-04)
Johannes Wohlrab, Reinhard H H Neubert, Marie-Luise Heskamp, Julia Michael
ZUSAMMENFASSUNG
Epicutaneous application of capsaicin causes a long-lasting analgesic effect by binding to the membrane transient receptor potential vanilloid 1 (TRPV1) on mechanoheat-sensitive C and Aδ fibres, changing axonal integrity and inhibiting neurogenic inflammatory processes. To date, no information is available regarding the cutaneous drug delivery of capsaicin following patch application. Using a Franz diffusion cell, the cutaneous concentration-time profiles 30, 60 and 90 min after application of a patch containing 8% capsaicin (640 µg/cm(2)) on ex vivo thin (mamma) and thick (plantar) human skin were investigated at 32 °C, and additionally at 42 °C for thin skin and 10 °C for thick skin. An HPLC-MS method was used for the analytic detection of capsaicin. The results show that already after a 30-min application of the 8% capsaicin patch, an equilibrium reservoir can be found in the stratum corneum in both thick and thin skin. Under physiological temperature conditions, a sufficient bioavailability of capsaicin in the cutaneous target compartments can be found. Raising the temperature to 42 °C has no relevant impact on the concentration-time profile, while reducing the temperature to 10°C leads to a significantly lower bioavailability. After 30 min of application, a sufficient cutaneous bioavailability of capsaicin is reached in thick as well as thin skin. Whether shorter application times may suffice to achieve therapeutic effectiveness requires further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Wasser, BioPerformance Certified
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Wasser, endotoxin, free
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O