Merck
  • Benzofuran analogues of amphetamine and methamphetamine: studies on the metabolism and toxicological analysis of 5-APB and 5-MAPB in urine and plasma using GC-MS and LC-(HR)-MS(n) techniques.
Benzofuran analogues of amphetamine and methamphetamine: studies on the metabolism and toxicological analysis of 5-APB and 5-MAPB in urine and plasma using GC-MS and LC-(HR)-MS(n) techniques.
Analytical and bioanalytical chemistry (2014-12-05)
Jessica Welter, Pierce Kavanagh, Markus R Meyer, Hans H Maurer
ZUSAMMENFASSUNG
5-APB (5-(2-aminopropyl)benzofuran) and its N-methyl derivative 5-MAPB (N-methyl-5-(2-aminopropyl)benzofuran) are analogues of amphetamine and methamphetamine, respectively, and belong to the so-called novel psychoactive substances (NPS). They were consumed as stimulants or entactogens with euphoric and empathogenic effects. Being controlled in some countries, both compounds should be covered by drug testing in clinical and forensic toxicology. Therefore, metabolism studies have been performed by working up rat urine samples after a high single dose of the corresponding NPS with solid-phase extraction without and after enzymatic conjugates cleavage. The phase I metabolites were separated and identified after acetylation by GC-MS and/or LC-HR-MS(n) and the phase II metabolites by LC-HR-MS(n). The main metabolite of 5-APB was 3-carboxymethyl-4-hydroxy amphetamine and the main metabolites of 5-MAPB were 5-APB (N-demethyl metabolite) and 3-carboxymethyl-4-hydroxy methamphetamine. The cytochrome P450 (CYP) isoenzymes involved in the 5-MAPB N-demethylation were CYP1A2, CYP2B6, CYP2C19, and CYP2D6, and according to the kinetic parameters, CYP2B6 was responsible for the main part of the total CYP-dependent clearance. An intake of a common users' dose of 5-APB or 5-MAPB could be confirmed in rat urine using the authors' GC-MS and the LC-MS(n) standard urine screening approaches with the corresponding parent drugs as major target. In authentic human urine samples after ingestion of unknown doses of 5-MAPB, both metabolites could also be detected besides the parent drug. The plasma concentrations determined in six clinical cases ranged from 5 to 124 μg/L for 5-MAPB and from 1 to 38 μg/L for its N-demethyl metabolite 5-APB.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%