Merck
  • Complement C1q-induced activation of β-catenin signalling causes hypertensive arterial remodelling.
Complement C1q-induced activation of β-catenin signalling causes hypertensive arterial remodelling.
Nature communications (2015-02-27)
Tomokazu Sumida, Atsuhiko T Naito, Seitaro Nomura, Akito Nakagawa, Tomoaki Higo, Akihito Hashimoto, Katsuki Okada, Taku Sakai, Masamichi Ito, Toshihiro Yamaguchi, Toru Oka, Hiroshi Akazawa, Jong-Kook Lee, Tohru Minamino, Stefan Offermanns, Tetsuo Noda, Marina Botto, Yoshio Kobayashi, Hiroyuki Morita, Ichiro Manabe, Toshio Nagai, Ichiro Shiojima, Issei Komuro
ZUSAMMENFASSUNG
Hypertension induces structural remodelling of arteries, which leads to arteriosclerosis and end-organ damage. Hyperplasia of vascular smooth muscle cells (VSMCs) and infiltration of immune cells are the hallmark of hypertensive arterial remodelling. However, the precise molecular mechanisms of arterial remodelling remain elusive. We have recently reported that complement C1q activates β-catenin signalling independent of Wnts. Here, we show a critical role of complement C1-induced activation of β-catenin signalling in hypertensive arterial remodelling. Activation of β-catenin and proliferation of VSMCs were observed after blood-pressure elevation, which were prevented by genetic and chemical inhibition of β-catenin signalling. Macrophage depletion and C1qa gene deletion attenuated the hypertension-induced β-catenin signalling, proliferation of VSMCs and pathological arterial remodelling. Our findings unveil the link between complement C1 and arterial remodelling and suggest that C1-induced activation of β-catenin signalling becomes a novel therapeutic target to prevent arteriosclerosis in patients with hypertension.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Anti-aktives β-Catenin(Anti-ABC)-Antikörper, Klon 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Hydralazin -hydrochlorid
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dichloromethylenediphosphonic acid disodium salt
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Hydralazin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard