Merck
  • Levels of polycyclic aromatic hydrocarbons in maternal serum and risk of neural tube defects in offspring.
Levels of polycyclic aromatic hydrocarbons in maternal serum and risk of neural tube defects in offspring.
Environmental science & technology (2014-12-10)
Bin Wang, Lei Jin, Aiguo Ren, Yue Yuan, Jufen Liu, Zhiwen Li, Le Zhang, Deqing Yi, Lin-Lin Wang, Yali Zhang, Xilong Wang, Shu Tao, Richard H Finnell
ZUSAMMENFASSUNG
Polycyclic aromatic hydrocarbons (PAHs) are ubiquitous environmental pollutants, and have been reported to be a risk factor for human neural tube defects (NTDs). We investigated the relationship between PAH concentrations in maternal serum and NTD risk in offspring using a case-control study design, and explored the link between PAH concentrations to household energy usage characteristics and life styles. One hundred and seventeen women who had NTD-affected pregnancies (cases) and 121 women who delivered healthy infants (controls) were recruited in Northern China. Maternal blood samples were collected at pregnancy termination or at delivery. Twenty-seven PAHs were measured by gas chromatography-mass spectrometry. The concentrations of 13 individual PAHs detected were significantly higher in the cases than in the controls. Clear dose-response relationships between concentrations of most individual PAHs and the risk of total NTDs or subtypes were observed, even when potential covariates were adjusted for. High-molecular-weight PAHs (H-PAHs) showed higher risk than low-molecular-weight PAHs (L-PAHs). No associations between PAH concentrations and indoor life styles and energy usage characteristics were observed. It was concluded that maternal exposure to PAHs was associated with an increased risk of NTDs, and H-PAHs overall posed a higher risk for NTDs than L-PAHs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Benzol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
6-Benzylaminopurin, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phenanthren, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
6-Benzylaminopurin, ReagentPlus®, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acenaphthen, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Fluorbiphenyl, 96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
2-Phenoxyethanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Phenoxy-ethanol, ≥90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acenaphthylin, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure, technical grade
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phenanthren, sublimed grade, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Phenanthren, analytical standard, for environmental analysis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Acenaphthen, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Pyroglutaminsäure-2-naphthylamid, ≥99% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Phenanthren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Acenaphthylin, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
6-Benzylaminopurin, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Acenaphthylin, 75%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Acenaphthen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.