Merck
  • Simvastatin inhibits the proliferation of A549 lung cancer cells through oxidative stress and up-regulation of SOD2.
Simvastatin inhibits the proliferation of A549 lung cancer cells through oxidative stress and up-regulation of SOD2.
Die Pharmazie (2014-08-28)
Yanjie Li, Jieqin Fu, Xue Yuan, Changhua Hu
ZUSAMMENFASSUNG
Beyond lipid-lowering effect, statins, the 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase competitively inhibitors, have recently attracted wide concern on their anticarcinogenic properties in various cancer cells. However, the molecular mechanisms by which statins inhibit cancer cell proliferation remain unclear. In this study, we investigated the effect of simvastatin on cells proliferation and oxidative stress in the A549 lung cancer cells and the mechanisms underlying. MTT assay revealed that simvastatin inhibited the proliferation of A549 cells in a concentration- and time-dependent manner. Treatment with 50 microM simvastatin for 48 h significantly increased reactive oxygen species (ROS) production and malondialdehyde (MDA), a lipid peroxidation production, and augmented the activity of total superoxide dismutase (SOD) and manganese SOD (SOD2). Moreover, western blotting analysis showed that simvastatin effectively up-regulated the SOD2 relative protein level. These findings suggested that simvastatin could inhibit the proliferation of A549 lung cells through oxidative stress and up-regulation of SOD2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
2′,7′-Dichlorfluoreszeindiacetat, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®