Merck
  • Cucurbitacin-D-induced CDK1 mRNA up-regulation causes proliferation arrest of a non-small cell lung carcinoma cell line (NSCLC-N6).
Cucurbitacin-D-induced CDK1 mRNA up-regulation causes proliferation arrest of a non-small cell lung carcinoma cell line (NSCLC-N6).
Anticancer research (2014-09-10)
Catherine Jacquot, Benedicte Rousseau, Delphine Carbonnelle, Ioanna Chinou, Marine Malleter, Christophe Tomasoni, Christos Roussakis
ZUSAMMENFASSUNG
Despite progress in chemotherapeutic agents, non-small cell lung cancers (NSCLC) still have a poor survival rate. Thus, development of new therapeutic strategies, specifically against cancer cells is still required. For this purpose, we treated the non-small cell lung cancer cell line NSCLC-N6 with the natural product cucurbitacin D (CucD) - extracted from the plant Ecballium elaterium in order first to assess its in vitro cytotoxicity, but also to study the genetic changes that it could bring out. CucD has shown a blocking in the G1 phase of the cell cycle in NSCLC-N6 cells prior to apoptotic cell death. The reverse transcriptase-polymerase chain reaction-differential display (RT-PCR-DD) technique was also applied on treated cells to elucidate the genetic mechanisms involved. We revealed an overexpression of Cyclin-dependent kinase 1 (CDK1) mRNA after treatment and, with the use of antisense oligonucleotides, an effective role in the proliferation arrest of NSCLC-N6 cells. The present study provides new insights about the mechanisms of proliferation arrest in tumor cells and open new ways of treatment to target tumor growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®