Merck
Multi-well microelectrode array recordings detect neuroactivity of ToxCast compounds.
Neurotoxicology (2014-07-06)
Pablo Valdivia, Matt Martin, William R LeFew, James Ross, Keith A Houck, Timothy J Shafer
ZUSAMMENFASSUNG
Spontaneous activity in neuronal cultures on microelectrode arrays (MEAs) is sensitive to effects of drugs, chemicals, and particles. Multi-well MEA (mwMEA) systems have increased throughput of MEAs, enabling their use for chemical screening. The present experiments examined a subset of EPA's ToxCast compounds for effects on spontaneous neuronal activity in primary cortical cultures using 48-well MEA plates. A first cohort of 68 compounds was selected from the ToxCast Phase I and II libraries; 37 were positive in one or more of 20 individual ToxCast Novascreen assays related to ion channels (NVS_IC), with the remainder selected based on known neuroactivity. A second cohort of 25 compounds was then tested with 20 originating from the ToxCast Phase I and II libraries (not hits in NVS_IC assays) and 5 known negatives from commercial vendors. Baseline activity (1h) was recorded prior to exposing the networks to compounds for 1h, and the weighted mean firing rate (wMFR) was determined in the absence and presence of each compound. Compounds that altered activity by greater than the weighted change of DMSO-treated wells plus 2SD were considered "hits". Of the first set of 68 compounds, 54 altered wMFR by more than the threshold, while in the second set, 13/25 compounds were hits. MEAs detected 30 of 37 (81.1%) compounds that were hits in NVS_IC assays, as well as detected known neurotoxicants that were negative in NVS_IC assays, primarily pyrethroids and GABAA receptor antagonists. Conversely, wMFR of cortical neuronal networks on MEAs was insensitive to nicotinic compounds, as only one neonicotinoid was detected by MEAs; this accounts for the bulk of non-concordant compounds between MEA and NVS_IC assays. These data demonstrate that mwMEAs can be used to screen chemicals efficiently for potential neurotoxicity, and that the results are concordant with predictions from ToxCast NVS_IC assays for interactions with ion channels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143091, Saccharin, traceable to primary standards (LGC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Saccharin, European Pharmacopoeia (EP) Reference Standard