Merck
  • Celecoxib exerts antitumor effects in canine mammary tumor cells via COX‑2‑independent mechanisms.
Celecoxib exerts antitumor effects in canine mammary tumor cells via COX‑2‑independent mechanisms.
International journal of oncology (2015-01-13)
Dai Tamura, Teruyoshi Saito, Kanae Murata, Masafumi Kawashima, Ryuji Asano
ZUSAMMENFASSUNG
Celecoxib plays antitumor roles via multiple mechanisms in a variety of human cancers. The aim of this study was to clarify the mechanism of action of celecoxib in canine mammary tumors. We examined the antitumor effects of celecoxib in AZACB canine mammary tumor cells expressing low levels of cyclooxygenase‑2 (COX‑2) to minimize the effect of COX‑2 on its activity. Our data revealed that celecoxib inhibited cell proliferation mainly via COX‑2‑independent mechanisms. Specifically, celecoxib decreased the proportion of cells in S phase and increased G2/M arrest, which was associated with increased expression of the cyclin‑dependent kinase inhibitors (CDKIs) p21 and p27. In addition, treatment with celecoxib downregulated COX‑2 expression, and induced apoptosis via both the intrinsic and extrinsic pathways. These findings suggest that celecoxib might be a useful agent for the treatment of canine mammary tumors, regardless of COX‑2 expression. In the future, it might be possible to use a combination of celecoxib and other antitumor agents to treat canine mammary tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
2,5-Dimethyl-Celecoxib, ≥98% (HPLC)
Sigma-Aldrich
Etodolac
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Dinoproston, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Etodolac, European Pharmacopoeia (EP) Reference Standard