Merck
  • Sex-specific effects of relaxin-3 on food intake and brain expression of corticotropin-releasing factor in rats.
Sex-specific effects of relaxin-3 on food intake and brain expression of corticotropin-releasing factor in rats.
Endocrinology (2014-11-19)
Christophe Lenglos, Juliane Calvez, Elena Timofeeva
ZUSAMMENFASSUNG
This study compared the effects of relaxin-3 (RLN3) on food intake, plasma corticosterone, and the expression of corticotropin-releasing factor (CRF) in male and female rats. RLN3 was injected into the lateral ventricle at 25, 200, and 800 pmol concentrations. RLN3 at 25 pmol increased food intake (grams) at 30 and 60 minutes after injection in female but not male rats. Female rats also showed higher increase in relative to body weight (BW) food intake (mg/g BW) for all RLN3 concentrations at 30 minutes and for 800 pmol of RLN3 at 60 minutes. Moreover, RLN3 at 800 pmol significantly increased 24-hour BW gain in female but not male rats. At 60 minutes after administration, 800 pmol of RLN3 produced a significant increase in plasma corticosterone and in the expression of CRF and c-fos mRNAs in the parvocellular paraventricular hypothalamic nucleus (PVN) in male but not female rats. The levels of c-fos mRNA in the magnocellular PVN were increased by RLN3 but did not differ between the sexes. Conversely, expression of CRF mRNA in the medial preoptic area was increased in female rats but was not sensitive to 800 pmol of RLN3. In the bed nucleus of the stria terminalis, 800 pmol of RLN3 significantly increased CRF mRNA expression in female but not male rats. Therefore, female rats showed more sensitivity and stronger food intake increase in response to RLN3. The differential effects of RLN3 on CRF expression in the PVN and bed nucleus of the stria terminalis may contribute to the sex-specific difference in the behavioral response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Corticosteron, ≥98.5% (HPLC)
Sigma-Aldrich
Corticosteron, ≥92%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methylenblau -Lösung, 0.05 wt. % in H2O
Sigma-Aldrich
Methylenblau -Lösung, suitable for microscopy
Sigma-Aldrich
Methylenblau -Lösung, suitable for microbiology
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methylenblaulösung 1,4 % (w/v) 95 % Ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Supelco
Corticosteron, VETRANAL®, analytical standard