Merck
  • Cloning, expression and functional characterization of in-house prepared human basic fibroblast growth factor.
Cloning, expression and functional characterization of in-house prepared human basic fibroblast growth factor.
Cell journal (2013-04-12)
Hassan Rassouli, Mohammad Sharif Tabe Bordbar, Mehran Rezaei Larijani, Mohammad Pakzad, Hossein Baharvand, Ghasem Hosseini Salekdeh
ZUSAMMENFASSUNG
Human basic fibroblast growth factor (bFGF) plays an important role in cellular proliferation, embryonic development, and angiogenesis as well as in several signaling pathways of various cell types. bFGF is an essential growth factor for the maintenance of undifferentiated human embryonic stem cells (hESCs) and human induced pluripotent stem cells (hiPSCs). In this experimental study, we present a straightforward method to produce biologically active recombinant human bFGF protein in E. coli that has long-term storage ability. This procedure provides a rapid, cost effective purification of a soluble human bFGF protein that is biologically active and functional as measured in hESCs and hiPSCs in vitro and in vivo. The results show no significant difference in function between our in-house produced and commercialized bFGF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Alkalische Leukozytenphosphatase-Kit, based on naphthol AS-BI and fast red violet LB
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
SAFC
Isopropyl β-D-1-Thiogalactopyranosid
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül) – FITC in Ziege hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-TRA-1-60-Antikörper, Klon TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anti-TRA-1-81-Antikörper, Klon TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Nanog in Maus hergestellte Antikörper, clone NNG-811, purified from hybridoma cell culture